Order Entry
United States
ContactUsLinkComponent
 
Anti-KIF3A Rabbit Polyclonal Antibody
Catalog # 10088-738
Supplier:  Proteintech
undefined
Anti-KIF3A Rabbit Polyclonal Antibody
Catalog # 10088-738
Supplier:  Proteintech
Supplier Number:  13930-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Kinesin Family member 3A
  • Antigen Symbol:
    KIF3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    FLA10|KLP-20
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10088-738
  • Supplier no.:
    13930-1-AP

Specifications

About this item

KIF3A is a subunit of Kinesin II that is essential for intraflagellar transport (IFT). It functions as the anterograde motor to deliver ciliary components to the tip where the axoneme is assembled. Disruption of KIF3A has been reported to block mammalian ciliogenesis. Recently KIF3A has been found to localize to the mother centriole and is required for centrioles organization. This antibody specifically recognizes the endogenous KIF3A protein in mouse embryonic fibroblasts (MEFs) but not in KIF3A-/- lysate. (23386061)

Western Blot: Human cerebellum Tissue, 1:200-1:2000; IHC: Human gliomas Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: KIF3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse