Order Entry
United States
ContactUsLinkComponent
Anti-CX3CR1 Rabbit Polyclonal Antibody
Anti-CX3CR1 Rabbit Polyclonal Antibody
Catalog # 10085-370
Supplier:  Proteintech
Anti-CX3CR1 Rabbit Polyclonal Antibody
Catalog # 10085-370
Supplier:  Proteintech
Supplier Number:  13885-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Chemokine (C-X3-C motif) receptor 1
  • Antigen Symbol:
    CX3CR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    CMKBRL1|V28|CCRL1|CMKDR1|GPR13|GPRV28
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10085-370
  • Supplier no.:
    13885-1-AP

Specifications

About this item

CX3CR1, also known as GPR13, V28 and fractalkine receptor, belongs to the 7-transmembrane G protein-coupled receptor (GPCR) family. It is expressed on microglia, astrocytes, NK cells, monocytes/macrophages, and a subpopulation of T cells. CX3CR1 is the receptor for fractalkine (CX3CL1) and mediates both its adhesive and migratory functions. It also acts as coreceptor with CD4 for HIV-1 virus envelope protein (in vitro), and some variations in the gene of CX3CR1 lead to increased susceptibility to HIV-1 infection and rapid progression to AIDS. Defects in CX3CR1 are a cause of susceptibility to age-related macular degeneration type 12 (ARMD12).

Western Blot: HL-60 Cells, 1:200-1:2000; IHC: Human tonsillitis Tissue, 1:20-1:200

Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: