To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Transcription Termination Factor RNA Polymerase 2
- Antigen Symbol:TTF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:ZGRF6|HuF2
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-476
- Supplier no.:13722-1-AP
Specifications
About this item
Transcription elongation is an key target for control of eukaryotic gene expression. After initiating from a promoter, the RNA polymerase II elongation complex is subjected to a block to elongation by the action of components of N-TEF (negative transcription elongation factor) . Transcription termination factor 2 (TTF2) is a DsDNA-dependent ATPase that acts as a transcription termination factor by coupling ATP hydrolysis with removal of RNA polymerase II from the DNA template. It also contribute to mitotic transcription repression, and involved in pre-mRNA splicing.
Western Blot:A2780 Cells, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200; IF: Hela, 1:20-1:200
Type: Primary
Antigen: TTF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat