Order Entry
United States
ContactUsLinkComponent
Anti-TULP3 Rabbit Polyclonal Antibody
Anti-TULP3 Rabbit Polyclonal Antibody
Catalog # 10096-506
Supplier:  Proteintech
undefined
Anti-TULP3 Rabbit Polyclonal Antibody
Catalog # 10096-506
Supplier:  Proteintech
Supplier Number:  13637-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tubby Like Protein 3
  • Antigen Symbol:
    TULP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    TUBL3
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10096-506
  • Supplier no.:
    13637-1-AP

Specifications

About this item

The tubby-like protein 3 (Tulp3) has been identified as a member of tubby and tubby-like proteins (TULPs) that plays an important role in maintenance and function of neuronal cells during development and post-differentiation. Tulp3 gene is essential for embryonic development in mice. Recently TULP3 has been reported to bind to the IFT(intraflagellar transport)-A complex and promote trafficking of G protein-coupled receptors into primary cilia,which is implicated in regulating neuronal function.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: TULP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat