Order Entry
ContactUsLinkComponent
Anti-EGR2 Rabbit Polyclonal Antibody
Anti-EGR2 Rabbit Polyclonal Antibody
Catalog # 10086-314
Supplier:  Proteintech
undefined
Anti-EGR2 Rabbit Polyclonal Antibody
Catalog # 10086-314
Supplier:  Proteintech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Early growth response protein 2
  • Antigen Symbol:
    EGR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    KROX20|Drosophila|homolog (early growth response-2)|KROX-20|Drosophila) |early growth response 2 (Krox-20 homolog|DKFZp686J1957|CMT4E|Krox-20 homolog|OTTHUMP00000062133 |CMT1D|EGR2|FLJ14547|early growth response 2 protein
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10086-314
  • Supplier no.:
    13491-1-AP

Specifications

About this item

Egr2 belongs to the Egr family that binds to DNA through highly conserved zinc finger domains and has a pivotal role in hindbrain development and myelination of the peripheral nervous system, development of T and/or B cells . It has been demonstrated that Egr2 is induced early upon TCR engagement in a NFAT-dependent manner, and that overexpression of Egr2 inhibits T cell activation. Overexpressing Egr2 shows an upregulation of E3-ligase Cbl-band reduced production of interleukin-2 (IL-2), suggesting that Egr2 is important for the maintenance of T cell tolerance by negatively regulating T cell activation.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: EGR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Frequently Bought Together