Anti-ETV6 Rabbit Polyclonal Antibody
10086-688
- Antibody Type:Primary
- Antigen Name:ETS variant 6
- Antigen Symbol:ETV6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:TEL/ABL|ETV6|ETS translocation variant 6|THC5|Transcription factor ETV6|TEL|TEL1|ETS-related protein Tel1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10086-688
- Supplier no.:13182-1-AP
Western Blot:A431 Cells, 1:500-1:5000
Type: Primary
Antigen: ETV6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat