Anti-SMARCA5 Rabbit Polyclonal Antibody
Catalog # 10094-854
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
- Antigen Symbol:SMARCA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:WCRF135|hISWI|SNF2H|ISWI|hSNF2H
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10094-854
- Supplier no.:13066-1-AP
Specifications
About this item
Western Blot: HepG2 Cells, 1:200-1:2000
Type: Primary
Antigen: SMARCA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat