Anti-FOXG1 Rabbit Polyclonal Antibody
Catalog # 10087-116
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Forkhead Box Protein G1
- Antigen Symbol:FOXG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Brain factor 2|KHL2|HBF-3|QIN|HBF-G2|FOXG1A|HFK1|HFK2|HFK3|FKHL4|Brain factor 1|FKHL3|Forkhead box protein G1|FKH2|BF2|FKHL1|HBF2|FOXG1B|BF1|FOXG1C|HBF-1|FKHL2
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10087-116
- Supplier no.:12764-1-AP
Specifications
About this item
Western Blot: Mouse Testis Tissue, 1:1000-1:10000
Type: Primary
Antigen: FOXG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat