Order Entry
United States
ContactUsLinkComponent
Anti-ICAM5 Rabbit Polyclonal Antibody
Anti-ICAM5 Rabbit Polyclonal Antibody
Catalog # 10088-556
Supplier:  Proteintech
undefined
Anti-ICAM5 Rabbit Polyclonal Antibody
Catalog # 10088-556
Supplier:  Proteintech
Supplier Number:  12759-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Intercellular Adhesion molecule 5 (telencephalin)
  • Antigen Symbol:
    ICAM5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    ICAM-3|TLN|Icam3|CD50|Tlcn
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10088-556
  • Supplier no.:
    12759-1-AP

Specifications

About this item

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: ICAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat