Order Entry
United States
ContactUsLinkComponent
Anti-TTP Rabbit Polyclonal Antibody
Anti-TTP Rabbit Polyclonal Antibody
Catalog # 10097-266
Supplier:  Proteintech
Anti-TTP Rabbit Polyclonal Antibody
Catalog # 10097-266
Supplier:  Proteintech
Supplier Number:  12737-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Tristetraprolin
  • Antigen Symbol:
    TTP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    RNF162A|NUP475|zfp-36|TTP|G0S24|GOS24|TIS11|ZFP36 ring finger protein
  • Storage Buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10097-266
  • Supplier no.:
    12737-1-AP

Specifications

About this item

The expression of many cytokines is regulated post-transcriptionally by factors that modulate mRNA transport, translation, and stability. Much of this regulation occurs by the binding and stabilizing, or destabilizing, of cytokine mRNAs by proteins that recognize adenosine and uridine-rich elements (AREs) in untranslated regions of target transcripts. Zfp36 is a mRNA-binding protein involved in post-transcriptional regulation of AU-rich element (ARE)-containing mRNAs. It was demonstrated to physically interact with the p65 subunit of nuclear factor-κB leading to decreased nuclear import and diminished transcriptional activation mediated by nuclear factor-κB. It acted by specifically binding ARE-containing mRNAs and promoting their degradation, and has a crucial role in the post-transcriptional regulation of tumor necrosis factor (TNF).

Western Blot: Mouse lung Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human breast cancer Tissue, 1:20-1:200

Type: Primary
Antigen: TTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat