Order Entry
United States
ContactUsLinkComponent
 
Anti-SULF2 Rabbit Polyclonal Antibody
  10094-598
Anti-SULF2 Rabbit Polyclonal Antibody
  10094-598
 :  12260-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Sulfatase 2
  • Antigen Symbol:
    SULF2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    KIAA1247|FLJ44488|MGC126411|HSULF-2|DKFZp313E091|RP5-1049G16.1|FLJ90554
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10094-598
  • Supplier no.:
    12260-1-AP

 

 

SULF2, also named as KIAA1247, belongs to the sulfatase family. It exhibits arylsulfatase activity and highly specific endoglucosamine-6-sulfatase activity. SULF2 can remove sulfate from the C-6 position of glucosamine within specific subregions of intact heparin. This protein is Glycosylation with MW about 130-150kd.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200

Type: Primary
Antigen: SULF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat