Anti-HuR Rabbit Polyclonal Antibody
Catalog # 10088-538
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:HuR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10088-538
- Supplier no.:11910-1-AP
Specifications
About this item
ELAVL1, also named as HUR, belongs to the RRM elav family. It is involved in 3'-UTR ARE-mediated MYC stabilization. ELAVL1 binds avidly to the AU-rich element in FOS and IL3/interleukin-3 mRNAs. In the case of the FOS AU-rich element, ELAVL1 binds to a core element of 27 nucleotides that contain AUUUA, AUUUUA and AUUUUUA motifs.
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:20-1:200; FC: MCF-7 Cells, N/A
Type: Primary
Antigen: HuR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Zebrafish