Order Entry
United States
ContactUsLinkComponent
Anti-CUX1 Rabbit Polyclonal Antibody
Anti-CUX1 Rabbit Polyclonal Antibody
  10085-362
undefined
Anti-CUX1 Rabbit Polyclonal Antibody
  10085-362
 :  11733-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Cut-like homeobox 1
  • Antigen Symbol:
    CUX1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    CDP/Cut|CUTL1|p75|CASP|p200|CUX|p100|GOLIM6|p110|Nbla10317|COY1|CDP|Clox|CDP1|Cux/CDP
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10085-362
  • Supplier no.:
    11733-1-AP

 

 

CUX1 is a transcription factors that regulates a large number of genes and microRNAs involved in multiple cellular processes, such as DNA replication, progression into S phase and later, the spindle assembly checkpoint that controls progression through mitosis. CUX1 acts by preventing binding of positively-acting CCAAT factors to promoters to mediated its gene reperssion. Also it's a component of nf-munr repressor, and binds to the matrix attachment regions (MARs) (5' and 3') of the immunoglobulin heavy chain enhancer. It can represses T-cell receptor (TCR) beta enhancer function by binding to MARbeta, an ATC-rich DNA sequence located upstream of the TCR beta enhancer

Western Blot: Human Brain Tissue, 1:1000-1:10000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CUX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: