Anti-CPI17 Rabbit Polyclonal Antibody
Catalog # 10085-110
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:CPI17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:Protein kinase C-potentiated inhibitor protein of 17 kDa|CPI17|PPP1INL|Protein phosphatase 1 regulatory subunit 14A|CPI-17|PPP1R14A|17 kDa PKC-potentiated inhibitory protein of PP1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10085-110
- Supplier no.:11658-1-AP
Specifications
About this item
Western Blot: HepG2 Cells, 1:1000-1:10000; IF:PC-3 Cells, 1:10-1:100
Type: Primary
Antigen: CPI17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: