Anti-IGF2BP2 Rabbit Polyclonal Antibody
10088-654
- Antibody Type:Primary
- Antigen Name:Insulin-like Growth Factor 2 mRNA binding Protein 1
- Antigen Symbol:IGF2BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:VICKZ2|IMP2|IMP-2
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10088-654
- Supplier no.:11601-1-AP
Insulin-like growth factor 2 mRNA-binding protein 2(IGF2BP2), ia a member of a family of mRNA binding protein, which can bind to several mammalian cells' mRNAs. Thus it may involve in the regulation of translation of mRNA. IGF2BP2's polymorphisms is risk factor for developing type 2 diabetes. This antibody raise against part of N-terminus of human IGF2BP2.
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IP: HEK-293 Cells, 1:200-1:2000
Type: Primary
Antigen: IGF2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat