Order Entry
United States
ContactUsLinkComponent
 
Anti-IFI30 Rabbit Polyclonal Antibody
  10088-592
undefined
Anti-IFI30 Rabbit Polyclonal Antibody
  10088-592
 :  11597-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Gamma-interferon-inducible Lysosomal Thiol Reductase
  • Antigen Symbol:
    IFI30
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    IFI-30|IP30|IP-30|GILT
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10088-592
  • Supplier no.:
    11597-1-AP

 

 

IFI30(interferon, gamma-inducible protein 30) is also named as IP30, GILT, IFI-30, MGC32056, IFI30 and belongs to the GILT family. It can facilitate the processing and presentation to antigen-specific T cells of protein antigens containing disulfide bonds. IFI30 is inducibly up-regulated by IFN-g, as are MHC class II molecules, the associated invariant chain, and HLA-DM, which catalyses peptide loading in MIICs and the 35 kDa species is the IFI30 proform, and the 30-kDa species is the mature form. It can exsit as a dimer.

Western Blot: Human Colon Tissue, 1:500-1:5000

Type: Primary
Antigen: IFI30
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human