Order Entry
United States
ContactUsLinkComponent
Anti-CAMKK2 Rabbit Polyclonal Antibody
Anti-CAMKK2 Rabbit Polyclonal Antibody
Catalog # 10084-038
Supplier:  Proteintech
undefined
Anti-CAMKK2 Rabbit Polyclonal Antibody
Catalog # 10084-038
Supplier:  Proteintech
Supplier Number:  11549-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Calcium/Calmodulin-dependent Protein Kinase 2 Beta
  • Antigen Symbol:
    CAMK2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Environmentally Preferable:
  • Antigen Synonyms:
    Calcium/calmodulin-dependent protein kinase kinase 2|KIAA0787|CaM-kinase kinase beta|CaM-KK beta|Calcium/calmodulin-dependent protein kinase kinase beta|CAMKK2|CaMKK beta|CAMKKB
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10084-038
  • Supplier no.:
    11549-1-AP

Specifications

About this item

CAMKK2(calcium/calmodulin-dependent protein kinase kinase 2) is also named as CAMKKB, KIAA0787 and belongs to the protein kinase superfamily. It regulates hypothalamic production of the orexigenic hormone NPY and contributes to the regulation of energy balance. CAMKK2 in vivo reduces tumor growth suggests that reactivation of CAMKK2 by development of new drugs may be a new strategy to prolong the period to respond androgen-deprivation therapy(ADT). It can be autophosphorylated. It has 7 isoforms produced by alternative splicing.

Western Blot:PC-3 Cells, 1:200-1:2000; IF:PC-3 Cells, 1:10-1:100

Type: Primary
Antigen: CAMKK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse