Anti-FBF1 Rabbit Polyclonal Antibody
Catalog # 10081-770
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Symbol:FBF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10081-770
- Supplier no.:11394-1-AP
Specifications
About this item
FBF1, also named as ALB and KIAA1863, is keratin-binding protein required for epithelial cell polarization. It is involved in apical junction complex (AJC) assembly via its interaction with PARD3. FBF1 is required for ciliogenesis. FBF1 has six isoforms with MW 125-127 kDa, 99 kDa, 69 kDa and 55 kDa. This antibody is induced by the fragment (492-792aa) of FBF1. It recognizes isoform 1, 2, 3, 5 and 6.
Western Blot: Mouse Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: FBF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat