Specifications
- Antibody Type:Primary
- Antigen Name:AE binding Protein 2
- Antigen Symbol:AEBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- ChIP:Yes
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10082-564
- Supplier no.:11232-2-AP
Specifications
About this item
AEBP2 belongs to the AEBP2/jing C2H2-type zinc-finger family. It is a DNA-binding transcriptional repressor. AEBP2 is a zinc finger protein that has been shown to interact with the mammalian Polycomb Repression Complex 2 (PRC2). It may interact with and stimulate the activity of the PRC2 complex, which methylates 'Lys-9' and 'Lys-27' residues of histone H3. AEBP2 is an evolutionarily well-conserved gene that is found in the animals ranging from flying insects to mammals. The antibody recognizes adult-specific larger form (52 kDa) and the embryo-specific smaller form (31 kDa).
Western Blot: Human Testis Tissue, 1:200-1:1000
Type: Primary
Antigen: AEBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat