Anti-ZNF24 Rabbit Polyclonal Antibody
Catalog # 10097-322
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Zinc Finger Protein 24
- Antigen Symbol:ZNF24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10097-322
- Supplier no.:11219-1-AP
Specifications
About this item
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human ovary tumor, 1:20-1:200
Type: Primary
Antigen: ZNF24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat