Specifications
- Antibody Type:Primary
- Antigen Name:Ras-related protein Rab-23
- Antigen Symbol:RAB23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10093-150
- Supplier no.:11101-1-AP
Specifications
About this item
Rab23 is a novel member of large Rab small GTPase family. It plays important roles during embryonic development. The brain-enriched Rab23 is the only Rab protein that is known to have a distinct function during central nervous system development, playing an essential role as a negative regulator of the Sonic Hedgehog (Shh) pathway. RAB23 has recently been identified in mesangial cells (MCs), and could serve as a biomarker that indicates the severity of FSGS ( focal segmental glomerulosclerosis). Mutations in RAB23 may result in ACPS2 ( acrocephalopolysyndactyly type 2).
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Liver cancer Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:500-1:5000
Type: Primary
Antigen: RAB23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat