To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Splicing Factor 3b Subunit 2, 145kDa
- Antigen Symbol:SF3B2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:SF3b1|Cus1|SAP145|SF3b150|SF3B145
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10094-436
- Supplier no.:10919-1-AP
Specifications
About this item
SF3B (splicing factor 3B) is a U2 snRNP-associated protein complex essential for spliceosome assembly. SF3B complex contains the spliceosome-associated protein (SAP) 49, 130, 145, and 155 has been demonstrated. And this complex is essential for U2 snRNP to anchor to the pre-mRNA. Splicing factor 3B subunit 2 (SF3B2), known as SAP145, forms complex with SAP49 which plays a role in cell cycle progression. SAP145, with predicated MW of 100 kDa, appears with MW of about 145 kDa which may be due to the post-translational modification.
- Western Blot: HEK-293 Cells, 1:200-1:1000; IHC: Human Colon cancer, 1:10-1:100
Type: Primary
Antigen: SF3B2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat