Order Entry
United States
ContactUsLinkComponent
 
Anti-FKBP52 Rabbit Polyclonal Antibody
  10082-136
undefined
Anti-FKBP52 Rabbit Polyclonal Antibody
  10082-136
 :  10655-1-AP-150UL

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    FKBP52
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping Temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. No.:
    10082-136
  • Supplier no.:
    10655-1-AP

 

 

FKBP52 (FK506 binding protein 4) is a member of the immunophilin protein family that comprises intracellular protein effectors of immunosuppressive drugs, and it is known as a steroid receptor-associated protein. As a novel regulator of microtubule dynamics, FKBP52 is associated with the motor protein dynein and with the cytoskeleton during mitosis. FKBP52 is highly expressed in CNS regions susceptible to Alzheimer's, and plays a role in modulating toxicity of Abeta peptides. The protein can associate with the Tau function, and may help to decipher and modulate the events involved in Tau-induced neurodegeneration. In addition, FKBP52 is likely to play a role in growth and development of the male genitalia.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100; IP: HeLa Cells, 1:500-1:5000; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: FKBP52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse