Anti-TRAPB Rabbit Polyclonal Antibody
10096-282
- Antibody Type:Primary
- Antigen Name:Translocon-associated Protein Subunit Beta
- Antigen Symbol:TRAPB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10096-282
- Supplier no.:10278-1-AP
TRAPB (translocon-associated protein subunit beta), also known as SSR2 (signal sequence receptor subunit beta), is a 22-kDa single-spanning membrane glycoprotein of the endoplasmic reticulum (ER) . It is a part of the tetrameric TRAP complex (TRAP-alpha, TRAP-beta, TRAP-delta and TRAP-gamma) which functions in regulating the retention of ER resident proteins . The TRAP complex is also involved in endoplasmic reticulum-associated degradation (ERAD) .
Western Blot: HepG2 Cells, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A; IF: HepG2 Cells, 1:10-1:100
Type: Primary
Antigen: TRAPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat