Anti-DHX38 Rabbit Polyclonal Antibody
Catalog # 10085-842
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Symbol:DHX38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- ChIP:Yes
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping Temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. No.:10085-842
- Supplier no.:10098-2-AP
Specifications
About this item
The protein encoded by this gene is a member of the DEAD/H box family of splicing factors, which are involved in pre-mRNA splicing. The yeast prp16 is an essential splicing factor that catalyzes RNA-dependent hydrolysis of nucleoside triphosphates, acting as an ATPase. This protein resembles yeast Prp16 more closely than other DEAD/H family members and also is a general splicing factor essential for catalytic step II during pre-mRNA processing. Western blot analysis showed it is a 140-KD nuclear protein expressed in various tissues.
Western Blot: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: DHX38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat