Order Entry
United States
ContactUsLinkComponent
Anti-IgY Goat Polyclonal Antibody (Biotin (NHS-LC))
Anti-IgY Goat Polyclonal Antibody (Biotin (NHS-LC))
  10149-604
undefined
Anti-IgY Goat Polyclonal Antibody (Biotin (NHS-LC))
  10149-604
 :  GTXCK-003-DBIO
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin Y
  • Antigen Symbol:
    IgY
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin (NHS-LC)
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgY
  • Reactivity:
    Chicken
  • Western Blot:
    Yes
  • Size:
    2.0 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Purified IgG
  • Form:
    Clear, colorless liquid
  • Storage Buffer:
    10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
  • Storage Temperature:
    2-8 °C
  • Concentration:
    1.5 mg/ml (E 1% at 280 nm = 13.0)
  • Shipping Temperature:
    2-8 °C
  • Immunogen:
    Chicken IgG/Y
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Affinity purified using solid phaseChicken IgG/Y
  • Cat. No.:
    10149-604
  • Supplier no.:
    IR1405

 

 

ImmunoReagents’ Biotinylated antibody conjugates are widely used due to their ability to enhance antibody signal in most immunoassays. Biotinylated secondary antibodies are used in most immunoassay formats (i.e., ELISA, Immunohistochemistry (IHC), and Western blotting). While Biotinylated antibodies by themselves do not produce a signal that can be visualized, biotin has a high affinity and specificity to Streptavidin/Avidin. Using ImmunoReagents’ streptavidin conjugates with biotinylated antibodies results in signal amplification.

  • Origin: US

ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

 : (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
 : Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: Biotin (NHS-LC)
Epitope:
Host: Goat
Isotype: IgY
Reactivity: Chicken