ContactUsLinkComponent
Giemsa's stain for microscopy, Sigma-Aldrich®
Irritant
Giemsa's stain for microscopy, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates
Irritant
Giemsa's stain for microscopy, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specification Test Results
  • Identity (UV/VIS-Spectrum):
    passes test
  • Loss on drying (105 °C):
    ≤ 10 %
  • Suitability for microscopy (Blood smear):
    passes test
  • Nuclei:
    violet
  • Lymphocytes (Plasma):
    Blue
  • Monocytes (Plasma):
    grey blue
  • basophilic granulocytes (Granules):
    dark violet
  • eosinophilic granulocytes (Granules):
    reddish to red-brown
  • Neutrophilic granulocytes (Granules):
    red-violet
  • Thrombocytes:
    violet
  • Erythrocytes:
    reddish-brownish
  • Blood parasites (Nuclei):
    lightred