Goat Serum, ImmunoReagents
About this item
ImmunoReagents’ normal goat serum is used as a blocking agent or as a control for immunoassays
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer. Our serum is supplied in lyophilized form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice, or to aliquot reconstituted serum and store frozen to prevent contamination.
Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your Avantor representative for more information.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
ImmunoReagents’ normal goat serum is used as a blocking agent or as a control for immunoassays
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.
All serum is obtained from healthy, disease free animals. Serum is lipid extracted and buffer exchanged against phosphate buffer. Our serum is supplied in lyophilized form without preservative. It is recommended to reconstitute the normal serum with DI water and add a preservative of choice, or to aliquot reconstituted serum and store frozen to prevent contamination.
Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your Avantor representative for more information.
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 10152-212
- 10152-210
- Application:
- ELISA, WB, IHC, IP FACS, ID and Labeling
- ELISA, WB, IHC, IP FACS, ID and Labeling
- Storage Temperature:
- Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.
- Store lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.
Specifications
Product Family Options
Product Information
- Serum formatAvailabilityPrice
- 10152-212Blocking and Control SeraSupplier Number: SP-004-VX2Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10152-212ApplicationELISA, WB, IHC, IP FACS, ID and LabelingStorage TemperatureStore lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria. - 10152-210Blocking and Control SeraSupplier Number: SP-004-VX10Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10152-210ApplicationELISA, WB, IHC, IP FACS, ID and LabelingStorage TemperatureStore lyophilized material at 2-8 °C. For long term storage after reconstitution, prepare small aliquots and store at -20 °C . For storage at 2-8 °C, add a preservative to prevent growth of bacteria.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



