

Trifluoroacetic acid 0.05% (v/v) in acetonitrile, LiChrosolv®, Supelco®
Catalog # EM4.80672.2500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM4.80672.2500
- Supplier no.:4.80672.2500
Specifications
About this item
:
:114.02 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:200-929-3
UN:1648
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169