Order Entry
United States
ContactUsLinkComponent
Trifluoroacetic acid 0.1% (v/v) in water, Supelco®
Trifluoroacetic acid 0.1% (v/v) in water, Supelco®
Catalog # EM4.80112.2500
Supplier:  MilliporeSigma
CAS Number:  
undefined
Trifluoroacetic acid 0.1% (v/v) in water, Supelco®
Catalog # EM4.80112.2500
Supplier:  MilliporeSigma
Supplier Number:  4.80112.2500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    2.5 L
  • Cat. No.:
    EM4.80112.2500
  • Supplier no.:
    4.80112.2500

Specifications

About this item

 :
 :114.02 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:200-929-3
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169