
1-Chlorobutane, LiChrosolv® for HPLC, Supelco®
EM1.01692.1000
:
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:EM1.01692.1000
- Supplier no.:1.01692.1000
:
:92.57 g/mol
:13,01559
:78.4 °C (1013 hPa)
:–123 °C
:0.8862 g/cm³ (20 °C)
:–17 °C
:Ambient
:203-696-6
:1127
:3,II
:M-Clarity™ quality level = MQ100
:GHS02
:109-69-3
:CH₃(CH₂)₃Cl
:MFCD00001009