Order Entry
United States
ContactUsLinkComponent
Toluene, EMPARTA® for analysis, Supelco®
Flammable
Health
Irritant
Toluene, EMPARTA® for analysis, Supelco®
Catalog # EM1.07019.4000
Supplier:  MilliporeSigma
CAS Number:  
undefined
Flammable
Health
Irritant
Toluene, EMPARTA® for analysis, Supelco®
Catalog # EM1.07019.4000
Supplier:  MilliporeSigma
Supplier Number:  1.07019.4000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    4 L
  • Cat. No.:
    EM1.07019.4000
  • Supplier no.:
    1.07019.4000

Specifications

About this item

 :
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
 :M-Clarity™ quality level = MQ200
REACH:05-2114615130-69
 :GHS02
 :GHS08
 :GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512