



n-Heptane ≥99%, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Catalog # EM1.04379.4000
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. No.:EM1.04379.4000
- Supplier no.:1.04379.4000
Specifications
Specification Test Results
- Purity (GC):≥ 99 %
- Identity (IR):conforms
- Color:≤ 10 Hazen
- Free acid (as CH₃COOH):≤ 0.0005 %
- Density (d 20 °C/20 °C):0.683 - 0.686
- Refractive index (n 20/D):1.387 - 1.388
- Boiling range (97°C-98°C):≥ 95 % (v/v)
- Aromatic hydrocarbons (as C₆H₆):≤ 0.1 %
- Sulfur compounds (as S):≤ 0.005 %
- Readily carbonizable substances:conforms
- Al (Aluminium):≤ 0.00001 %
- B (Boron):≤ 0.000002 %
- Ba (Barium):≤ 0.00001 %
- Ca (Calcium):≤ 0.00005 %
- Cd (Cadmium):≤ 0.000005 %
- Co (Cobalt):≤ 0.000002 %
- Cr (Chromium):≤ 0.000002 %
- Cu (Copper):≤ 0.000002 %
- Fe (Iron):≤ 0.00001 %
- Mg (Magnesium):≤ 0.00001 %
- Mn (Manganese):≤ 0.000002 %
- Ni (Nickel):≤ 0.000002 %
- Pb (Lead):≤ 0.00001 %
- Sn (Tin):≤ 0.00001 %
- Zn (Zinc):≤ 0.00001 %
- Evaporation residue:≤ 0.001 %
- Water:≤ 0.01 %
Specification Test Results
About this item
:
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:M-Clarity™ quality level = MQ300
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544