Order Entry
United States
Orders LinkContactUsLinkComponent
Chloroform, GR for analysis, Supelco®

Chloroform, GR for analysis, Supelco®

Toxic
Health
Supplier:   MilliporeSigma
Chloroform, GR for analysis, Supelco®
Supplier:  MilliporeSigma
Certificates
Chloroform, GR for analysis, Supelco®
Chloroform, GR for analysis, Supelco®

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :M-Clarity™ quality level = MQ300
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:05-2114614906-45

Documents

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $140.82
    $140.82
    Log in for availability
    Case of 6 (1l)
    Case of 6 (1l) - $844.92
    $844.92
    Each (1l)
       $140.82

    Case of 6 (1l)
       $844.92

    Supplier Number: 1024421000
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Glass bottle
    2.5 L
    Log in for availability
    Log in for availability
    Log in for availability
    Each (2,500ml)
    Each (2,500ml) - $286.08
    $286.08
    Log in for availability
    Case of 4 (2,500,ml)
    Case of 4 (2,500,ml) - $1,144.32
    $1,144.32
    Each (2,500ml)
       $286.08

    Case of 4 (2,500,ml)
       $1,144.32

    Supplier Number: 1024422500
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.