

Dichloromethane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
UN:1593
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:200-838-9
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:20 g/l (20 °C)
- Melting Point:-95 °C
- Molar Mass:84.93 g/mol
- Boiling Point:40 °C (1013 hPa)
- Vapor Pressure:475 hPa (20 °C)
- Refractive Index:1.42
- Explosion Limit:13 - 22 %(V)
- Density:1.33 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:605 °C DIN 51794
- Synonyms:DCM, Methylene chloride, Methylene dichloride
- Hill Formula:CH2Cl2
- Chemical Formula:CH2Cl2
- LD50 oral:LD50 Rat 1600 mg/kg
- LD50 dermal:LD50 Rat greater than 2000 mg/kg
- Storage:Store at +15°C to +25°C
Specification Test Results
Frequently Bought Together