Acetone, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
Boiling Pt:56.2
Melting Pt:-95.4
Density:0.792 g/cm³ (20 °C)
Molecular Weight:58.08 g/mol
Flash Pt:<–20 °C (closed cup)
EINECS:200-662-2
UN:1090
ADR:3 II
Merck Index:13,00067
:M-Clarity™ quality level = MQ100
REACH:01-2119471330-49
:GHS02
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-95.4 °C
- Molar Mass:58.08 g/mol
- Boiling Point:56.2 °C (1013 hPa)
- Vapor Pressure:233 hPa (20 °C)
- Refractive Index:1.35868 (20 °C)
- Explosion Limit:2.6 - 12.8 %(V)
- Density:0.79 g/cm3 (20 °C)
- pH:5 - 6 (395 g/l, H2O, 20 °C)
- Ignition Point:465 °C DIN 51794
- Synonyms:Dimethyl ketone, Propanone, 2-Propanone
- Hill Formula:C3H6O
- Chemical Formula:CH3COCH3
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Solvents in 5L Aluminum Bottles
Product Literature
ghs_1.00020
SDS
Frequently Bought Together