Orders LinkContactUsLinkComponent
VWR® Vortex Mixers
VWR® Vortex Mixers
   Sustainable Options Available
Supplier:   VWR International
VWR® Vortex Mixers
Supplier:  VWR International
   Sustainable Options Available
Certificates
VWR® Vortex Mixers
6 Options Available Below
Product Details & Documents
Fixed high speed mixing and variable speed models.
  • Five year warranty for parts and labor
  • All models include a cup head and three inch platform
  • Manufactured in the USA
VWR® Fixed Speed Vortex Mixers start mixing when the cup head is depressed. Speed is a full 3200 rpm to provide vigorous mixing of samples.
VWR® Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.
VWR® Digital Vortex Mixers are ideal for applications that demand repeatable results. Microprocessor control maintains set speed for strong, consistent mixing action. Touch pad controls and independent LED displays for speed and time. Timer will display elapsed time or when programmed to a user defined time limit, the unit will shut off when time reaches zero.Two modes of operation; continuous mode when using the optional accessory attachments, or touch mode which activates mixing when depressing the cup head.
VWR® Pulsing Vortex Mixers are ideal for glass bead cell disruption/homogenization and provide a powerful pulsing vortex action.Capable of complete cell disruption of samples in only minutes.Unique pulsing action mixes at maximum speed for 30 seconds and rests for 10 seconds reducing heat generation while providing more effective mixing and disruption. Displayed time counts up during continuous operation and counts down during timed runs. Also includes a stainless steel microtube holder with built-in cup head.
Certifications: TUV listed. RoHS compliant.
Ordering information: All models include a cup head, three-inch head with cover, and a 72" (183 cm), 3-wire cord and plug. Pulsing Vortex Mixer includes a stainless steel microtube holder with built-in cup head and holds up to (12) 1.5 to 2 ml microtubes.
Vortex Mixers with NIST traceable calibration certificate provided by an ISO/IEC 17025 accredited laboratory. Multiple data points within the speed range and time function are reported on the certificate with the associated uncertainties.
Specifications for Product Family
Specifications
  • Catalog No:
  • 10153-834
  • 10153-838
  • 10153-842
  • 10153-812
  • 10153-816
  • 10153-820
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
  • Shipping Weight:
  • 10 lbs.
  • 10 lbs.
  • 10 lbs.
  • 10 lbs.
  • 10 lbs.
  • 10 lbs.
  • L×W×H:
  • 7×4.4×5.3"
  • 7×4.4×5.3"
  • 7×4.4×5.3"
  • 7×4.4×5.3"
  • 7×4.4×5.3"
  • 7×4.4×5.3"
  • Duty Rating:
  • Intermittent Duty
  • Intermittent Duty
  • Intermittent Duty
  • Intermittent Duty
  • Intermittent Duty
  • Intermittent Duty
  • Environmental Certification:
  • RoHS
  • RoHS
  • RoHS
  • RoHS
  • RoHS
  • RoHS
Specifications
Product Family Options
Vortex Mixers
Product Information
  • Electrical
    Orbit
    Speed
    Availability
    Price
  • 10153-834
    VWR® Fixed Speed Vortex Mixer
    VWR® Fixed Speed Vortex Mixer
    120 V, 50/60 Hz, 1.2 A, 135 W
    4.9 mm
    3200 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $608.63
    $608.63
    New Customer Promotional Price
    Each
       $608.63
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-834
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
  • 10153-838
    VWR® Analog Vortex Mixer
    VWR® Analog Vortex Mixer
    120 V, 50/60 Hz, 1.2 A, 135 W
    4.9 mm
    300 - 3200 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $631.84
    $631.84
    New Customer Promotional Price
    Each
       $631.84
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-838
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
  • 10153-842
    VWR® Digital Vortex Mixer
    VWR® Digital Vortex Mixer
    120 V, 50/60 Hz, 1.2 A, 135 W
    4.9 mm
    500 - 3000 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $776.76
    $776.76
    New Customer Promotional Price
    Each
       $776.76
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-842
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
  • 10153-812
    VWR® Pulsing Vortex Mixer
    VWR® Pulsing Vortex Mixer
    120 V, 50/60 Hz, 1.2 A, 135 W
    2.5 mm
    500 - 3000 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $948.93
    $948.93
    New Customer Promotional Price
    Each
       $948.93
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-812
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
Vortex Mixers with NIST Traceable Certificates
Product Information
  • Electrical
    Orbit
    Speed
    Availability
    Price
  • 10153-816
    VWR® Digital Vortex Mixer with NIST Certificate
    VWR® Digital Vortex Mixer with NIST Certificate
    120 V, 50/60 Hz, 1.2 A, 135 W
    4.9 mm
    500 - 3000 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $710.16
    $710.16
    New Customer Promotional Price
    Each
       $710.16
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-816
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
  • 10153-820
    VWR® Pulsing Vortex Mixer with NIST Certificate
    VWR® Pulsing Vortex Mixer with NIST Certificate
    120 V, 50/60 Hz, 1.2 A, 135 W
    2.5 mm
    500 - 3000 rpm
    Log in for availability
    Log in for availability
    Each
    Each - $1,154.97
    $1,154.97
    New Customer Promotional Price
    Each
       $1,154.97
    Specifications:
    Product available on GSA Advantage®
      Waste Reducing
    More
    Specifications
    Cat. No.
    10153-820
    Environmentally Preferable
    Waste Reducing
    Shipping Weight
    10 lbs.
    L×W×H
    7×4.4×5.3"
    Duty Rating
    Intermittent Duty
    Environmental Certification
    RoHS
    Product available on GSA Advantage®
      Waste Reducing
Accessories for tubes and plates
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Microtube and microplate holder kit with insert retainer
    Log in for availability
    Log in for availability
    Each
    Each - $151.97
    $151.97
    Each
       $151.97
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Tube holder, 9 to 13 mm
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $41.74
    $41.74
    Pack of 2
       $41.74
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Insert retainer
    Log in for availability
    Log in for availability
    Each
    Each - $187.14
    $187.14
    Each
       $187.14
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Microtube holder
    Log in for availability
    Log in for availability
    Each
    Each - $90.04
    $90.04
    Each
       $90.04
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Tube holder, 20 to 25 mm
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $41.99
    $41.99
    Pack of 2
       $41.99
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Microplate holder
    Log in for availability
    Log in for availability
    Each
    Each - $84.40
    $84.40
    Each
       $84.40
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Flat foam insert, 160 mm (6.3") diameter
    Log in for availability
    Log in for availability
    Each
    Each - $41.46
    $41.46
    Each
       $41.46
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Tube holder kit with insert retainer
    Log in for availability
    Log in for availability
    Each
    Each - $264.86
    $264.86
    Each
       $264.86
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Tube holder, 14 to 19 mm
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $41.99
    $41.99
    Pack of 2
       $41.99
Other accessories
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Stainless steel microtube holder
    Log in for availability
    Log in for availability
    Each
    Each - $433.90
    $433.90
    Each
       $433.90
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Cup head
    Log in for availability
    Log in for availability
    Each
    Each - $57.21
    $57.21
    Each
       $57.21
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Vessel harness
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $12.73
    $12.73
    Pack of 2
       $12.73
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Single tube holder
    Log in for availability
    Log in for availability
    Each
    Each - $245.60
    $245.60
    Each
       $245.60
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Head, 76 mm (3")
    Log in for availability
    Log in for availability
    Each
    Each - $55.30
    $55.30
    Each
       $55.30
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Ampule holder, 10 to 17 mm
    Log in for availability
    Log in for availability
    Each
    Each - $274.16
    $274.16
    Each
       $274.16
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Rubber head cover, 76 mm (3")
    Log in for availability
    Log in for availability
    Each
    Each - $62.72
    $62.72
    Each
       $62.72
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Single tube holder accessory for 18×1.5 - 2 ml tubes
    Log in for availability
    Log in for availability
    Each
    Each - $120.37
    $120.37
    Each
       $120.37
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Ampule holder, 15 to 17 mm
    Log in for availability
    Log in for availability
    Each
    Each - $184.07
    $184.07
    Each
       $184.07
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Head with rubber cover, 76 mm (3")
    Log in for availability
    Log in for availability
    Each
    Each - $82.78
    $82.78
    Each
       $82.78
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Single tube holder adapter for Vortex Genie
    Log in for availability
    Log in for availability
    Each
    Each - $121.95
    $121.95
    Each
       $121.95
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Single tube holder accessory for 24×0.5 ml tubes
    Log in for availability
    Log in for availability
    Each
    Each - $117.00
    $117.00
    Each
       $117.00
Grinding beads
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 200 µm
    200 µm
    Log in for availability
    Log in for availability
    Each
    Each - $478.23
    $478.23
    Each
       $478.23
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 800 µm
    800 µm
    Log in for availability
    Log in for availability
    Each
    Each - $256.41
    $256.41
    Each
       $256.41
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia low protein binding grinding beads, 800 µm
    800 µm
    Log in for availability
    Log in for availability
    Each
    Each - $426.46
    $426.46
    Each
       $426.46
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 200 µm
    200 µm
    Log in for availability
    Log in for availability
    Each
    Each - $360.34
    $360.34
    Each
       $360.34
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia low protein binding grinding beads, 400 µm
    400 µm
    Log in for availability
    Log in for availability
    Each
    Each - $426.46
    $426.46
    Each
       $426.46
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 400 µm
    400 µm
    Log in for availability
    Log in for availability
    Each
    Each - $259.21
    $259.21
    Each
       $259.21
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 400 µm
    400 µm
    Log in for availability
    Log in for availability
    Each
    Each - $339.69
    $339.69
    Each
       $339.69
Product Information
  • Catalog #
    Description
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 800 µm
    800 µm
    Log in for availability
    Log in for availability
    Each
    Each - $339.69
    $339.69
    Each
       $339.69
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cup head
    Cup head
    58816-156
    Log in for availability
    Each - $57.21
Product Information
  • Catalog #
    Availability
    Price
  • Rubber head cover, 76 mm (3")
    Rubber head cover, 76 mm (3")
    58816-154
    Log in for availability
    Each - $62.72
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder, 20 to 25 mm
    Tube holder, 20 to 25 mm
    58816-142
    Log in for availability
    Pack of 2 - $41.99
Product Information
  • Catalog #
    Availability
    Price
  • Head with rubber cover, 76 mm (3")
    Head with rubber cover, 76 mm (3")
    12620-872
    Log in for availability
    Each - $82.78
Product Information
  • Catalog #
    Availability
    Price
  • Flat foam insert, 160 mm (6.3") diameter
    Flat foam insert, 160 mm (6.3") diameter
    12620-884
    Log in for availability
    Each - $41.46
Product Information
  • Catalog #
    Availability
    Price
  • Microplate holder
    Microplate holder
    12620-878
    Log in for availability
    Each - $84.40
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 200 µm
    Zirconia acid washed grinding beads, 200 µm
    12621-158
    200 µm
    Log in for availability
    Each - $360.34
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 200 µm
    Zirconia molecular biology grade grinding beads, 200 µm
    12621-160
    200 µm
    Log in for availability
    Each - $478.23
Product Information
  • Catalog #
    Availability
    Price
  • Single tube holder adapter for Vortex Genie
    Single tube holder adapter for Vortex Genie
    12621-252
    Log in for availability
    Each - $121.95
Product Information
  • Catalog #
    Availability
    Price
  • Vessel harness
    Vessel harness
    58816-136
    Log in for availability
    Pack of 2 - $12.73
Product Information
  • Catalog #
    Availability
    Price
  • Ampule holder, 15 to 17 mm
    Ampule holder, 15 to 17 mm
    12620-886
    Log in for availability
    Each - $184.07
Product Information
  • Catalog #
    Availability
    Price
  • Ampule holder, 10 to 17 mm
    Ampule holder, 10 to 17 mm
    12620-888
    Log in for availability
    Each - $274.16
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel microtube holder
    Stainless steel microtube holder
    12620-890
    Log in for availability
    Each - $433.90
Product Information
  • Catalog #
    Availability
    Price
  • Single tube holder accessory for 24×0.5 ml tubes
    Single tube holder accessory for 24×0.5 ml tubes
    12620-894
    Log in for availability
    Each - $117.00
Product Information
  • Catalog #
    Availability
    Price
  • Single tube holder
    Single tube holder
    12620-898
    Log in for availability
    Each - $245.60
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 800 µm
    Zirconia acid washed grinding beads, 800 µm
    12621-146
    800 µm
    Log in for availability
    Each - $256.41
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 800 µm
    Zirconia molecular biology grade grinding beads, 800 µm
    12621-148
    800 µm
    Log in for availability
    Each - $339.69
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia low protein binding grinding beads, 400 µm
    Zirconia low protein binding grinding beads, 400 µm
    12621-156
    400 µm
    Log in for availability
    Each - $426.46
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia low protein binding grinding beads, 800 µm
    Zirconia low protein binding grinding beads, 800 µm
    12621-150
    800 µm
    Log in for availability
    Each - $426.46
Product Information
  • Catalog #
    Availability
    Price
  • Microtube holder
    Microtube holder
    12620-876
    Log in for availability
    Each - $90.04
Product Information
  • Catalog #
    Availability
    Price
  • Microtube and microplate holder kit with insert retainer
    Microtube and microplate holder kit with insert retainer
    12620-874
    Log in for availability
    Each - $151.97
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia acid washed grinding beads, 400 µm
    Zirconia acid washed grinding beads, 400 µm
    12621-152
    400 µm
    Log in for availability
    Each - $259.21
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Zirconia molecular biology grade grinding beads, 400 µm
    Zirconia molecular biology grade grinding beads, 400 µm
    12621-154
    400 µm
    Log in for availability
    Each - $339.69
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder, 9 to 13 mm
    Tube holder, 9 to 13 mm
    58816-138
    Log in for availability
    Pack of 2 - $41.74
Product Information
  • Catalog #
    Availability
    Price
  • Insert retainer
    Insert retainer
    58816-132
    Log in for availability
    Each - $187.14
Product Information
  • Catalog #
    Availability
    Price
  • Single tube holder accessory for 18×1.5 - 2 ml tubes
    Single tube holder accessory for 18×1.5 - 2 ml tubes
    12620-896
    Log in for availability
    Each - $120.37
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder kit with insert retainer
    Tube holder kit with insert retainer
    58816-130
    Log in for availability
    Each - $264.86
Product Information
  • Catalog #
    Availability
    Price
  • Head, 76 mm (3")
    Head, 76 mm (3")
    58816-152
    Log in for availability
    Each - $55.30
Product Information
  • Catalog #
    Availability
    Price
  • Tube holder, 14 to 19 mm
    Tube holder, 14 to 19 mm
    58816-140
    Log in for availability
    Pack of 2 - $41.99