Polyacetal Spherical Joint Clamp, Kimble Chase
Catalog # KT675310-0013
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Color:Violet
- Cat. No.:KT675310-0013
- Joint size:Fits 12/5, 12/312/2 (mm)
- Supplier no.:675310-0013
Specifications
About this item
An easy to use Spherical Joint positive closure clamp made of tough, color coded plastic for use with ball and socket joints
Clamp will not scratch glass and can be autoclaved to 200°C. This clamp has good chemical resistance at a lower cost than metal clips.