ContactUsLinkComponent
Whatman™ Nuclepore Track-Etched Polycarbonate Gold-Coated Membranes, Whatman products (Cytiva)
Whatman™ Nuclepore Track-Etched Polycarbonate Gold-Coated Membranes, Whatman products (Cytiva)
Catalog # 10035-596
Supplier:  Avantor
undefined
Whatman™ Nuclepore Track-Etched Polycarbonate Gold-Coated Membranes, Whatman products (Cytiva)
Catalog # 10035-596
Supplier:  Avantor
About this item
Nuclepore track-etched polycarbonate gold-coated membrane.
  • Low protein binding and low extractables ensuring no sample contamination
  • High chemical resistance and good thermal stability for a wide range of samples
  • Low, consistent ash and low tare weights
  • Smooth flat surface for good visibility of particles
Nuclepore track-etched polycarbonate membranes are manufactured from high-quality polycarbonate film and have sharply defined pore sizes, high flow rates, and excellent chemical and thermal resistance. The membranes have a smooth flat surface and exhibit very low levels of extractables.