Mini-UniPrep G2 is an all-in-one syringeless filter with a borosilicate glass chamber. It replaces syringe filters for autosampler-supported UHPLC/HPLC sample preparation with easier handling and fewer consumable components.
- Features an integrated autosampler vial, plunger with attached filter membrane, and septum/cap
- Borosilicate glass vials eliminate plastic-based leachables that can originate from polypropylene vials
- Designed to hold up to 0.5 ml of unfiltered sample that can then be loaded directly into an autosampler
- Easy to see when a sample has been filtered
- Wide range of membranes with 0.2 and 0.45 µm pore sizes to meet specific requirements
- Also available in amber glass for light-sensitive samples
A variety of filter membrane media are available to meet your needs.
Mini-UniPrep Syringeless Filters combine a membrane filter, autosampler vial, septum, and plunger into a single device to provide efficient sample filtration for Ultra High Performance Liquid Chromatography (UHPLC)/High Performance Liquid Chromatography (HPLC) analysis.
Once compressed, Mini-Uniprep G2 can be placed into any autosampler that accepts a 12 mm × 32 autosampler vial for injection into your instrument. Alternatively, the septum can be pierced with a needle and the sample drawn off for manual injection into an analyzer.
Simplifying UHPLC/HPLC sample preparation minimizes instrument downtime resulting from use of unfiltered samples.
Browse the Whatman line to find the right filter and compressor for your application.
GE Healthcare Environmental testing for water air and soil Brochure (CAN) GE Healthcare Environmental testing for water air and soil Brochure (US) GE Healthcare Environmental Testing for Water Brochure GE Healthcare Environmental Testing for Water Brochure (CA) GE_Healthcare_Whatman_Filtration_Products_for_Pharmaceutical_Quality_Control_(US)