ContactUsLinkComponent
KIMBLE® Thermometer Inlet Adapter with Hose Connection, DWK Life Sciences
KIMBLE® Thermometer Inlet Adapter with Hose Connection, DWK Life Sciences
Catalog # KT197000-2440
undefined
KIMBLE® Thermometer Inlet Adapter with Hose Connection, DWK Life Sciences
Catalog # KT197000-2440
Supplier Number:  197000-2440
Specifications
  • [ST] joint:
    24/40
  • Cat. No.:
    KT197000-2440
  • Overall height:
    155 mm
  • Fits tubing I.D.:
    ⅜"
  • Supplier no.:
    197000-2440
Specifications
About this item
Adapter features a hose connection on the side.
  • Side hose connection
  • Accommodates thermometer
  • Manufactured from borosilicate glass
The hose connection on the side accepts 3/8” ID flexible tubing.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.