ContactUsLinkComponent
Syringe Needle Flask Adapters, Kimble Chase
Syringe Needle Flask Adapters, Kimble Chase
Catalog # KT197800-2440
undefined
Syringe Needle Flask Adapters, Kimble Chase
Catalog # KT197800-2440
Supplier Number:  197800-2440
Specifications
  • Cap type:
    Blind hole rubber stopper
  • Cat. No.:
    KT197800-2440
  • [ST] joint size:
    [ST] 24/40
  • Supplier no.:
    197800-2440
Specifications
About this item
Provides access to flasks and other containers when pierced by a syringe needle
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
When used in conjunction with standard syringes, liquid samples may be added or withdrawn from multiple neck flasks. Allows syringe needles to be used as capillary bleed tubes or bubblers.
Ordering information: Supplied with one Blind Hole Rubber Stopper fitted to the top.