Order Entry
United States
ContactUsLinkComponent
KIMBLE® Bleed Type Inlet Adapter with Dual Hose Connections and Standard Taper Joint, DWK Life Sciences
KIMBLE® Bleed Type Inlet Adapter with Dual Hose Connections and Standard Taper Joint, DWK Life Sciences
Catalog # KT181000-2440
undefined
KIMBLE® Bleed Type Inlet Adapter with Dual Hose Connections and Standard Taper Joint, DWK Life Sciences
Catalog # KT181000-2440
Supplier Number:  181000-2440

Specifications

  • Description:
    Adapter Gas Inlet, Tube Length 200 mm
  • Cat. No.:
    KT181000-2440
  • Overall height:
    355 mm
  • Fits tubing I.D.:
    ⅜"
  • Supplier no.:
    181000-2440

Specifications

About this item

Designed with two side hose connectors for venting or introducing gas/vacuum into a system.

  • Two side hose connectors
  • [ST] joint
  • Manufactured from borosilicate glass

The hose connections fit 3/8” ID flexible tubing.

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.