Order Entry
United States
ContactUsLinkComponent
 
Agarose LE, Ultrapure for molecular biology, for electrophoresis
 
undefined
Agarose LE, Ultrapure for molecular biology, for electrophoresis
 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Form:
    Granular, free-flowing powder
  • Assay (unspecified) Gel Background:
    Passes test
  • Assay (unspecified) Gel Point (1.5%):
    36 ± 1.5°C
  • Assay (unspecified) Gel Strength (1%):
    ≥ 1200 gm/cm2
  • Assay (unspecified) Gel Strength (1.5%):
    ≥ 2500 gm/cm2
  • Comment: EEO (-Mr):
    0.05 - 0.13
  • Comment: Turbidity (1.5%):
    ≤ 40 Np
  • DNase:
    Passes test
  • Residual water:
    ≤ 7%
  • Residue on ignition:
    ≤ 0.4%
  • Impurity content: Sulfate (SO4):
    ≤ 0.14%
  • Melting Point: Remelt Point (1.5%):
    88 ± 1.5°C
  • RNase:
    Passes test

 

Frequently Bought Together