Order Entry
United States
ContactUsLinkComponent
KIMBLE® Offset Connecting Adapters, DWK Life Sciences
KIMBLE® Offset Connecting Adapters, DWK Life Sciences
Catalog # KT188000-2440
undefined
KIMBLE® Offset Connecting Adapters, DWK Life Sciences
Catalog # KT188000-2440
Supplier Number:  188000-2440

Specifications

  • [ST] joint:
    24/40
  • Cat. No.:
    KT188000-2440
  • Overall length:
    180 mm
  • Offset:
    40 mm
  • Supplier no.:
    188000-2440

Specifications

About this item

The offset connecting adapter is used with multiple entry flasks or reaction vessels.

  • Offset design
  • Joints are the same size
  • Manufactured from borosilicate glass

Design helps avoid possible contact with stirrer shafts

Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Joints are the same size.