Order Entry
United States
ContactUsLinkComponent
 
QBSF Serum Free Media, Quality Biological
    
 
undefined
QBSF Serum Free Media, Quality Biological
    

 

  • Environmentally Preferable:
    Sustainable Packaging
  • Sterility:
    Sterile filtered
  • Environmental Certification:
    ECO LOGOS
  • Shipping Conditions:
    Ambient temperature
  • pH:
    7.0 ±0.2

 

Frequently Bought Together