Riser Shelf for RTW Table, Production Basics
Supplier: Production Basics
Certificates
About this item
Mounts to the top of the RTW Table to provide extra workspace and privacy for your lab technician
- Finished with rounded corners and protective T-mold edge
For use with any Production Basics RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench.
Metal shelf frame with standard laminate surface.
Includes 1 laminate shelf, 1 riser frame, 1 parts and hardware packet.
Ordering information: When purchased at same time as RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench the colors will be made to match. If purchasing at separate time, specify Riser Frame and Laminate colors otherwise will be default Blue Frame/White Laminate. Frame available in Light Neutral/Almond, Blue, White, Grey and Black. Worksurface available in White, Grey and Beige. Accessory is ready to mount on a RTW Lab Bench, PedSys Lab Bench or Easy Lift 4-post Lab Bench of same width.
Frequently Bought Together