Wire Shelf, Production Basics
Supplier: Production Basics
Certificates
About this item
Chrome shelf provides a clean, technical look and allows ambient light to pass through to worksurface.
- Weight capacity is 34.1 kg (75 lbs.)
For use with on any C-Leg Lab Bench. Add uprights to mount accessories on RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench, Basic Organizer or Easy Lift 4-post Lab Bench.
Includes 1 wire shelf, 1 shelf brackets set, 1 parts and hardware packet.
Ordering information: Chrome shelf is supported by brackets finished in black wrinkle epoxy powder coat finish. Other standard frame color for brackets (Light Neutral/Almond, Blue, White, or Grey) available by request. Accessory is ready to mount on a C-Leg Lab Bench; or otherwise must purchase Uprights, sold separately, if mounting on a RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench or Easy Lift 4-post Lab Bench.
Frequently Bought Together