Accessories for RTW Adjustable - Height Worktables, Production Basics
Add mobility to a Production Basics RTW Lab Bench
For use with any RTW Lab Bench Lab Bench.
Includes (4) four 10.2 diameter cm (4 in.) diameter thermoplastic casters with swivel locks and brakes. This total-lock feature ensures a rigid laboratory workstation.
: Add mobility to you RTW Table. Includes four (4) 4" thermoplastic grey casters with swivel locks and brakes. This total lock feature ensure a rigid, stable workstation, while making it easy to re-position. Surface height is fixed at 35.5". If order for existing RTW bench, specify the Frame color to match. Frame colors available in Light Neutral/Almond, Blue, White, Grey and Black.
Frequently Bought Together