Adjustable Leg for C - Leg Lab Bench, Production Basics
Supplier: Production Basics
Certificates
About this item
C-leg for stand- alone workstations (two required per stand alone) or to support the end stations in a multi-station configuration.
- Durable workstation legs feature a rugged 14-gauge steel design for maximum support
- Provides clearance beneath matching depth worksurface
For onsite retro-fit or replacement on any Production Basics C-leg lab bench.
Ordering information: Order leg replacement same as worksurface depth. Specify a frame color otherwise will be default blue frame. Frame available in light neutral/almond, blue, white, grey and black.
Delivery: Includes 1 adjustable C-leg, 4 leveling feet, 1 parts and hardware packet.
Frequently Bought Together